Novus Biologicals
Manufacturer Code:NBP192646
Catalog # NBP192646
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | This antibody was developed against Recombinant Protein corresponding to amino acids:RINTVQYGTSDNKKDNTLQCGMEVNSSILSGCASNHPKNSAQNLTKDSTLKHEGFYSENLTESRKFLKSREKQSSLTEIKGSVYETTYSPPECPFCGKIEEHSEDMETHVKTKHANLLDIPLEDCDQPLYDCPMC |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: C6orf113 chromosome 6 open reading frame 113 dJ412I7.3 zinc finger with UFM1-specific peptidase domain zinc finger with UFM1-specific peptidase domain protein; DUB; Lys-63-specific deubiquitinase ZUFSP; Zinc finger with UFM1-specific peptidase domain protein; Zinc finger-containing ubiquitin peptidase 1
Gene Aliases: C6orf113; dJ412I7.3; ZUFSP; ZUP1
UniProt ID: (Human) Q96AP4
Entrez Gene ID: (Human) 221302
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.