Invitrogen
Manufacturer Code:PA569969
Catalog # PIPA569969
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | synthetic peptide directed towards the N terminal region of human ZSWIM3 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: C20orf164; protein phosphatase 1, regulatory subunit 174; Zinc finger SWIM domain-containing protein 3; zinc finger, SWIM domain containing 3; zinc finger, SWIM-type containing 3
Gene Aliases: 4921517A06Rik; C20orf164; C86566; PPP1R174; ZSWIM3
UniProt ID: (Human) Q96MP5, (Mouse) Q8CFL8
Entrez Gene ID: (Rabbit) 100339103, (Human) 140831, (Pig) 100152744, (Dog) 485901, (Horse) 100056419, (Rat) 311630, (Mouse) 67538, (Guinea pig) 100719405
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.