Invitrogen
Manufacturer Code:PA566332
Catalog # PIPA566332
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit Human ZSWIM3 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: C20orf164; protein phosphatase 1, regulatory subunit 174; Zinc finger SWIM domain-containing protein 3; zinc finger, SWIM domain containing 3; zinc finger, SWIM-type containing 3
Gene Aliases: C20orf164; PPP1R174; ZSWIM3
UniProt ID: (Human) Q96MP5
Entrez Gene ID: (Human) 140831
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.