Novus Biologicals
Manufacturer Code:NBP221056
Catalog # NBP221056
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant protein encompassing a sequence within the center region of human ZSWIM2. The exact sequence is proprietary. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: centromere/kinetochore protein zw10 homolog HZW10 KNTC1AP MGC149821 zeste white 10 homolog ZW10 (Drosophila) homolog centromere/kinetochore protein ZW10 homolog centromere/kinetochore protein ZW10 kinetochore associated homolog (Drosophila); E3 ubiquitin-protein ligase ZSWIM2; MEKK1-related protein X; MEX; RING-type E3 ubiquitin transferase ZSWIM2; Zinc finger SWIM domain-containing protein 2; zinc finger, SWIM domain containing 2; zinc finger, SWIM-type containing 2; ZZ-type zinc finger-containing protein 2
Gene Aliases: MEX; ZSWIM2; ZZZ2
UniProt ID: (Human) Q8NEG5
Entrez Gene ID: (Human) 151112
Molecular Function:
ubiquitin-protein ligase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.