Abnova
Manufacturer Code:H00151112K
Catalog # 89-200-711
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Immunogen | A synthetic peptide of human ZSWIM2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Rabbit monoclonal antibody raised against a human ZSWIM2 peptide using ARM Technology.
For Research Use Only
Protein Aliases: E3 ubiquitin-protein ligase ZSWIM2; MEKK1-related protein X; MEX; MGC33890 ZZZ2; RING-type E3 ubiquitin transferase ZSWIM2; Zinc finger SWIM domain-containing protein 2; zinc finger, SWIM domain containing 2; zinc finger, SWIM-type containing 2; ZZ-type zinc finger-containing protein 2
Gene Aliases: MEX; ZSWIM2; ZZZ2
UniProt ID: (Human) Q8NEG5
Entrez Gene ID: (Human) 151112
Molecular Function: ubiquitin-protein ligase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.