Novus Biologicals
Manufacturer Code:NBP29409120UL
Catalog # NB022742
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 411-485 of human ZSWIM1 (NP_542170.3). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°:C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: FLJ10036 FLJ16343 homolog of Drosophila Zwilch hZwilch KNTC1AP MGC111034 protein zwilch homolog Zwilch kinetochore associated homolog (Drosophila); Zinc finger SWIM domain-containing protein 1; zinc finger, SWIM domain containing 1; zinc finger, SWIM-type containing 1
Gene Aliases: C20orf162; ZSWIM1
UniProt ID: (Human) Q9BR11
Entrez Gene ID: (Human) 90204
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.