Invitrogen
Manufacturer Code:PA541174
Catalog # PIPA541174
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the middle region of human ZSCAN5 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: zinc finger and SCAN domain containing 5; Zinc finger and SCAN domain containing 5A ZNF495 ZSCAN5; Zinc finger and SCAN domain-containing protein 5A; Zinc finger protein 495
Gene Aliases: ZNF495; ZSCAN5; ZSCAN5A
UniProt ID: (Human) Q9BUG6
Entrez Gene ID: (Human) 79149
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.