Invitrogen
Manufacturer Code:PA5110610
Catalog # PIPA5110610
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide of mouse ZSCAN4. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Gm397 XM_142517 Zinc finger and SCAN domain containing protein 4C Zscan4c Zscan4d; Zinc finger and SCAN domain containing protein 4C
Gene Aliases: Gm397; XM_142517; Zscan4c; Zscan4d
UniProt ID: (Mouse) Q80VJ6
Entrez Gene ID: (Mouse) 245109
Molecular Function:
nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.