Invitrogen
Manufacturer Code:PA5112907
Catalog # PIPA5112907
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant Human U2 small nuclear ribonucleoprotein auxiliary factor 35 kDa subunit-related protein 2 protein (191-372AA) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 35kDa 5031411E02Rik A230052C13Rik C77286 CCCH type zinc finger RNA-binding motif and serine/arginine rich protein 2 LOC100351607 Renal carcinoma antigen NY-REN-20 RGD1559763 U2 small nuclear ribonucleoprotein auxiliary factor (U2AF) 35 kDa related sequence 2 U2 small nuclear ribonucleoprotein auxiliary factor 35 kDa subunit-related protein 2 U2 small nuclear ribonucleoprotein auxiliary factor small subunit 2 U2 small nuclear RNA auxiliary factor 1-like 2 U2(RNU2) small nuclear RNA auxiliary factor 1-like 2 U2AF1L2 U2AF1RS2 U2AF1-RS2 U2AF35-related protein URP ZC3H22 zinc finger (CCCH type) RNA binding motif and serine/arginine rich 2 zinc finger (CCCH type) RNA-binding motif and serine/arginine rich 2 zinc finger CCCH-type RNA binding motif and serine/arginine rich 2 ZRSR2; CCCH type zinc finger, RNA-binding motif and serine/arginine rich protein 2; Renal carcinoma antigen NY-REN-20; U2 small nuclear ribonucleoprotein auxiliary factor (U2AF), 35 kDa, related sequence 2; U2 small nuclear ribonucleoprotein auxiliary factor 35 kDa subunit-related protein 2; U2 small nuclear ribonucleoprotein auxiliary factor, small subunit 2; U2(RNU2) small nuclear RNA auxiliary factor 1-like 2; U2AF35-related protein; URP; zinc finger (CCCH type), RNA binding motif and serine/arginine rich 2; zinc finger (CCCH type), RNA-binding motif and serine/arginine rich 2
Gene Aliases: 35kDa; 5031411E02Rik; A230052C13Rik; C77286; U2AF1-RS2; U2AF1L2; U2AF1RS2; URP; ZC3H22; ZRSR2
UniProt ID: (Human) Q15696
Entrez Gene ID: (Human) 8233, (Mouse) 22184
Molecular Function:
RNA binding protein
nucleic acid binding
ribonucleoprotein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.