Novus Biologicals
Manufacturer Code:NBP157307
Catalog # NBP157307
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptides corresponding to ZRSR2(zinc finger (CCCH type) RNA-binding motif and serine/arginine rich 2) The peptide sequence was selected from the N terminal of ZRSR2 (NP_005080). Peptide sequence LKEQWEEQQRKEREEEEQKRQEKKEKEEALQKMLDQAENELENGT |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CCCH type zinc finger RNA-binding motif and serine/arginine rich protein 2 Renal carcinoma antigen NY-REN-20 U2 small nuclear ribonucleoprotein auxiliary factor 35 kDa subunit-relatedprotein 2 U2 small nuclear ribonucleoprotein auxiliary factor small subunit 2 U2(RNU2) small nuclear RNA auxiliary factor 1-like 2MGC142014 U2AF1L2 U2AF1RS2MGC142040 U2AF1-RS2U2AF35-related protein URPU2 small nuclear RNA auxiliary factor 1-like 2 zinc finger (CCCH type) RNA-binding motif and serine/arginine rich 2; CCCH type zinc finger, RNA-binding motif and serine/arginine rich protein 2; Renal carcinoma antigen NY-REN-20; U2 small nuclear ribonucleoprotein auxiliary factor 35 kDa subunit-related protein 2; U2 small nuclear ribonucleoprotein auxiliary factor, small subunit 2; U2(RNU2) small nuclear RNA auxiliary factor 1-like 2; U2AF35-related protein; URP; zinc finger (CCCH type), RNA binding motif and serine/arginine rich 2; zinc finger (CCCH type), RNA-binding motif and serine/arginine rich 2
Gene Aliases: U2AF1-RS2; U2AF1L2; U2AF1RS2; URP; ZC3H22; ZRSR2
UniProt ID: (Human) Q15696
Entrez Gene ID: (Human) 8233
Molecular Function: RNA binding protein nucleic acid binding ribonucleoprotein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.