Invitrogen
Manufacturer Code:PA570488
Catalog # PIPA570488
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | synthetic peptide directed towards the C-terminal region of human ZRANB3 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 4933425L19Rik AH2 DNA annealing helicase and endonuclease ZRANB3 annealing helicase 2 zinc finger Ran-binding domain-containing protein 3 zinc finger RAN-binding domain containing 3; AH2; Annealing helicase 2; DNA annealing helicase and endonuclease ZRANB3; DNA annealing helicase ZRANB3; Endonuclease ZRANB3; Zinc finger Ran-binding domain-containing protein 3; zinc finger, RAN-binding domain containing 3
Gene Aliases: 4933425L19Rik; AH2; AI316834; C730006D09; RGD1564952; ZRANB3
UniProt ID: (Human) Q5FWF4, (Mouse) Q6NZP1
Entrez Gene ID: (Rabbit) 100350819, (Horse) 100050746, (Pig) 100157618, (Dog) 476128, (Human) 84083, (Rat) 304761, (Mouse) 226409
Molecular Function: DNA binding protein DNA helicase helicase nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.