Invitrogen
Manufacturer Code:PA526829
Catalog # PIPA526829
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 665-692 amino acids from the C-terminal region of human ZRANB1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: hTrabid; TRABID TRAF-binding protein domain hTrabid ubiquitin thioesterase ZRANB1 zinc finger Ran-binding domain-containing protein 1; TRAF-binding domain-containing protein; TRAF-binding protein; TRAF-binding protein domain; Ubiquitin thioesterase ZRANB1; Zinc finger Ran-binding domain-containing protein 1; zinc finger, RAN-binding domain containing 1
Gene Aliases: 9330160G10Rik; D7Wsu87e; Gm1956; TRABID; ZRANB1
UniProt ID: (Human) Q9UGI0, (Mouse) Q7M760
Entrez Gene ID: (Human) 54764, (Mouse) 360216
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.