Invitrogen
Manufacturer Code:PA541470
Catalog # PIPA541470
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the N-terminal of human ZNF259 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Zinc finger protein 259; Zinc finger protein 259 ZPR1; Zinc finger protein ZPR1
Gene Aliases: AI303781; fc17g10; RGD1562173; si:rp71-56i13.4; wu:fc17g10; Zfp259; zgc:56478; ZNF259; ZPR1
UniProt ID: (Human) O75312, (Bovine) Q2TBX0, (Mouse) Q62384
Entrez Gene ID: (Dog) 479428, (Human) 8882, (Rabbit) 100328795, (Bovine) 504523, (Horse) 100062470, (Rat) 500989, (Mouse) 22687, (Guinea pig) 100729662, (Zebrafish) 406382
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.