Invitrogen
Manufacturer Code:PA532173
Catalog # PIPA532173
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein fragment corresponding to a region within amino acids 400 and 888 of Mouse MAP3K12 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: DLK; DLK MUK Zpk MAPK-upstream kinase dual leucine zipper bearing kinase leucine-zipper protein kinase mitogen activated protein kinase kinase kinase 12 mixed lineage kinase zipper (leucine) protein kinase; Dual leucine zipper bearing kinase; dual leucine zipper kinase; dual leucine zipper kinase DLK; leucine zipper protein kinase; Leucine-zipper protein kinase; MAPK-upstream kinase; Mitogen activated protein kinase 12 (Zipper (leucine) protein kinase); mitogen activated protein kinase kinase kinase 12; Mitogen-activated protein kinase kinase kinase 12; Mixed lineage kinase; MUK; protein kinase MUK; Zipper (leucine) protein kinase; ZPK
Gene Aliases: DLK; MAP3K12; MEKK12; MUK; PK; ZPK; ZPKP1
UniProt ID: (Human) B3KSS9, (Rat) Q63796, (Mouse) Q60700
Entrez Gene ID: (Human) 7786, (Rat) 25579, (Mouse) 26404
Molecular Function:
kinase
non-receptor serine/threonine protein kinase
non-receptor tyrosine protein kinase
protein kinase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.