Invitrogen
Manufacturer Code:PA5100731
Catalog # PIPA5100731
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human MAP3K12 corresponding to a region within N-terminal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: DLK; DLK Dual leucine zipper bearing kinase dual leucine zipper kinase dual leucine zipper kinase DLK leucine zipper protein kinase leucine-zipper protein kinase M3KC Map3k12 MAPK-upstream kinase MEKK12 Mitogen activated protein kinase 12 (Zipper (leucine) protein kinase) mitogen activated protein kinase kinase kinase 12 mitogen-activated protein kinase kinase kinase 12 Mixed lineage kinase MUK PK protein kinase MUK zipper (leucine) protein kinase ZPK ZPKP1; Dual leucine zipper bearing kinase; dual leucine zipper kinase; dual leucine zipper kinase DLK; leucine zipper protein kinase; Leucine-zipper protein kinase; MAPK-upstream kinase; Mitogen activated protein kinase 12 (Zipper (leucine) protein kinase); mitogen activated protein kinase kinase kinase 12; Mitogen-activated protein kinase kinase kinase 12; Mixed lineage kinase; MUK; protein kinase MUK; Zipper (leucine) protein kinase; ZPK
Gene Aliases: DLK; MAP3K12; MEKK12; MUK; PK; ZPK; ZPKP1
UniProt ID: (Human) B3KSS9, (Rat) Q63796, (Mouse) Q60700
Entrez Gene ID: (Human) 7786, (Rat) 25579, (Mouse) 26404
Molecular Function:
kinase
non-receptor serine/threonine protein kinase
non-receptor tyrosine protein kinase
protein kinase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.