Novus Biologicals
Manufacturer Code:NBP157768
Catalog # NBP157768
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptides corresponding to ZPBP2 (zona pellucida binding protein 2) The peptide sequence was selected from the middle region of ZPBP2)(50ug). Peptide sequence VRLDSCRPGFGKNERLHSNCASCCVVCSPATFSPDVNVTCQTCVSVLTYG. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CAAX prenyl protease 1 homolog EC 3.4.24.84 FACE1FLJ14968 FACE-1zinc metalloproteinase (STE24 homolog yeast) Farnesylated proteins-converting enzyme 1 farnesylated-proteins converting enzyme 1 HGPS Hutchinson-Gilford progeria syndrome MADB Prenyl protein-specific endoprotease 1 PRO1 Ste24p STE24zinc metallopeptidase (STE24 homolog yeast) zinc metallopeptidase (STE24 homolog S. cerevisiae) Zinc metalloproteinase Ste24 homolog; Zona pellucida-binding protein 2; ZPBP-like protein
Gene Aliases: ZPBP2; ZPBPL
UniProt ID: (Human) Q6X784
Entrez Gene ID: (Human) 124626
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.