Novus Biologicals
Manufacturer Code:NBP294587100UL
Catalog # NB023698
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to a sequence within amino acids 400-500 of human ZP4 (NP_067009.1). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°:C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Processed zona pellucida sperm-binding protein 4; zinc finger and homeodomain protein 1 zinc fingers and homeobox 1 zinc fingers and homeoboxes 1 zinc fingers and homeoboxes protein 1 zinc-fingers and homeoboxes 1; Zona pellucida glycoprotein 4; Zona pellucida protein B; Zona pellucida sperm-binding protein 4; Zp-4
Gene Aliases: ZBP; Zp-4; ZP1; ZP4; ZPB
UniProt ID: (Human) Q12836
Entrez Gene ID: (Human) 57829
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.