Novus Biologicals
Manufacturer Code:NBP2865660.1ML
Catalog # NB034190
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | The immunogen is a synthetic peptide directed towards the C-terminal region of human ZP3 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. Aliquot and store at -20°:C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Processed zona pellucida sperm-binding protein 3; Sperm receptor; Zona pellucida glycoprotein 3; zona pellucida glycoprotein 3 (sperm receptor); zona pellucida glycoprotein 3A (sperm receptor); zona pellucida glycoprotein 3B; zona pellucida glycoprotein ZP3; Zona pellucida protein C; Zona pellucida sperm-binding protein 3; Zp-3; ZP3A/ZP3B
Gene Aliases: Zp-3; ZP3; ZP3A; ZP3B; ZPC
UniProt ID: (Human) P21754
Entrez Gene ID: (Human) 7784
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.