Invitrogen
Manufacturer Code:PA542736
Catalog # PIPA542736
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the C-terminal of human ZP2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 75-kDa rabbit zona pellucida protein; Processed zona pellucida sperm-binding protein 2; zona pellucida 2 glycoprotein; zona pellucida A protein; Zona pellucida glycoprotein 2; zona pellucida glycoprotein 2 (sperm receptor); Zona pellucida glycoprotein 2 (sperm receptor) ZPA Zp-2; zona pellucida glycoprotein ZP2; Zona pellucida protein A; Zona pellucida sperm-binding protein 2; Zp-2
Gene Aliases: Zp-2; ZP2; ZPA
UniProt ID: (Dog) P47983, (Human) Q05996, (Bovine) Q9BH10, (Rat) O54767
Entrez Gene ID: (Dog) 403988, (Human) 7783, (Rabbit) 100009069, (Bovine) 280963, (Horse) 100059065, (Guinea pig) 100713508, (Rat) 81828
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.