Invitrogen
Manufacturer Code:PA530879
Catalog # PIPA530879
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fragment corresponding to a region within amino acids 1 and 313 of Human ZP2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Processed zona pellucida sperm-binding protein 2; Zona pellucida glycoprotein 2; zona pellucida glycoprotein 2 (sperm receptor); zona pellucida glycoprotein ZP2; Zona pellucida protein A; Zona pellucida sperm-binding protein 2; Zp-2; ZPA Zp-2 zona pellucida glycoprotein ZP2 zona pellucida protein A zona pellucida sperm-binding protein 2
Gene Aliases: Zp-2; ZP2; ZPA
UniProt ID: (Human) Q05996
Entrez Gene ID: (Human) 7783
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.