Invitrogen
Manufacturer Code:PA585619
Catalog # PIPA585619
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Carrier-protein conjugated synthetic peptide encompassing a sequence within the C-terminus region of human CSDA. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: cold shock domain protein A; Cold shock domain-containing protein A; DNA-binding protein A; Muscle Y-box protein YB2; RYB-A; Y-box binding protein 3 Antibody YBX3 Antibody CSDA1 Antibody ZONAB Antibody DBPA Antibody; Y-box protein 3; Y-box-binding protein 3; Y-box-binding protein A
Gene Aliases: Csda; Dbpa; Dpba; MSY3; MSY4; oxyR; Yb2; Ybx3
UniProt ID: (Rat) Q62764, (Mouse) Q9JKB3
Entrez Gene ID: (Rat) 83807, (Mouse) 56449
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.