Invitrogen
Manufacturer Code:PA535151
Catalog # PIPA535151
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 152-181 amino acids from the central region of human CSDA |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: cold shock domain protein A; Cold shock domain-containing protein A; cold-shock domain containing A1; cold-shock domain protein A; CSDA DBPA DNA-binding protein A Cold shock domain-containing protein A Single-strand DNA-binding protein NF-GMB; DNA-binding protein A; Single-strand DNA-binding protein NF-GMB; Y box binding protein 3; Y-box protein 3; Y-box-binding protein 3; ZO-1-associated nucleic acid-binding protein
Gene Aliases: CSDA; CSDA1; DBPA; Dpba; MSY3; MSY4; oxyR; Yb2; YBX3; ZONAB
UniProt ID: (Human) P16989, (Mouse) Q9JKB3
Entrez Gene ID: (Human) 8531, (Mouse) 56449
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.