Invitrogen
Manufacturer Code:PA517155
Catalog # PIPA517155
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide corresponding to the carboxy-terminal sequence of mouse ZO-2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Friedreich ataxia region gene X104 (tight junction protein ZO-2); Friedreich ataxia region gene X104 (tight junction protein ZO-2) tight junction protein ZO 2 tight junction protein ZO-2 zona occludens protein 2 zonula occludens protein 2; Tight junction protein 2; tight junction protein 2 (zona occludens 2); Tight junction protein ZO-2; Tight junction protein ZO-2 (Zonula occludens 2 protein) (Zona occludens 2 protein) (Tight junction protein 2); zona occludens 2; Zona occludens protein 2; Zonula occludens protein 2
Gene Aliases: C9DUPq21.11; DFNA51; DUP9q21.11; PFIC4; TJP2; X104; ZO-2; ZO2
UniProt ID: (Dog) Q95168, (Human) Q9UDY2, (Mouse) Q9Z0U1
Entrez Gene ID: (Bovine) 407101, (Dog) 403854, (Human) 9414, (Mouse) 21873, (Rat) 115769
Molecular Function:
cell junction protein
tight junction
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.