Proteintech
Manufacturer Code:217731AP150UL
Catalog # 50-560-891
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | ZO-1 Fusion Protein Ag16454 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Tight junction protein 1; tight junction protein 1 (zona occludens 1); Tight junction protein 1 Tight junction protein ZO 1 TJP1 ZO 1 ZO1 ZO-1 Zona occludens protein 1 Zonula occludens protein 1; Tight junction protein ZO-1; zona occludens 1; Zona occludens protein 1; zonula occludens 1 protein; Zonula occludens protein 1
Gene Aliases: I79_005485; TJP1; ZO-1; ZO1
UniProt ID: (Dog) O97758, (Human) Q07157, (Mouse) P39447
Entrez Gene ID: (Dog) 403752, (Human) 7082, (Mouse) 21872, (Rat) 292994, (Chinese hamster) 100773110
Molecular Function:
cell junction protein
tight junction
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.