Novus Biologicals
Manufacturer Code:NBP190758
Catalog # NBP190758
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | This antibody was developed against Recombinant Protein corresponding to amino acids:PNSSTSEVGLEASPGAAPDLRRTWKGGHELPSCACCCEPQPSPAGPSAGAAGSSTLFLGPHLYEGSGPAGGEPQSGSSQGLYGLHPDHLPRTDGVKYEGLPCCFYEEKQVAR |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CTA-292E10.6; E3 ubiquitin-protein ligase ZNRF3; novel C3HC4 type Zinc finger (ring finger); novel C3HC4 type Zinc finger (ring finger) zinc and ring finger 3 zinc/RING finger protein 3; RING finger protein 203; RING-type E3 ubiquitin transferase ZNRF3; Zinc/RING finger protein 3
Gene Aliases: BK747E2.3; KIAA1133; RNF203; ZNRF3
UniProt ID: (Human) Q9ULT6
Entrez Gene ID: (Human) 84133
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.