Invitrogen
Manufacturer Code:PA5116330
Catalog # PIPA5116330
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 40-67 amino acids from the N-terminal region of human ZNF93 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: HPF34 HTF34 TF34 Zinc finger protein 505 zinc finger protein 93 Zinc finger protein HTF34 ZNF505 ZNF93; Zinc finger protein 505; Zinc finger protein 93; Zinc finger protein HTF34
Gene Aliases: HPF34; HTF34; TF34; ZNF505; ZNF93
UniProt ID: (Human) P35789
Entrez Gene ID: (Human) 81931
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.