Invitrogen
Manufacturer Code:PA541197
Catalog # PIPA541197
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the middle region of human CCDC16 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: coiled-coil domain containing 16; Coiled-coil domain-containing protein 16; hypothetical protein MGC20398; orphan maintenance of genome 1; Ovus mutant candidate gene 1 protein; Zinc finger protein 830; Zinc finger protein 830-like protein
Gene Aliases: 2410003C20Rik; AV301118; AW048207; CCDC16; OMCG1; Zfp830; ZNF830
UniProt ID: (Human) Q96NB3, (Rat) Q3MHS2, (Mouse) Q8R1N0
Entrez Gene ID: (Human) 91603, (Bovine) 539497, (Horse) 100071676, (Dog) 480608, (Rat) 497967, (Mouse) 66983, (Guinea pig) 100715797
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.