Novus Biologicals
Manufacturer Code:NBP188402
Catalog # NBP188402
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | This antibody was developed against Recombinant Protein corresponding to amino acids:YYHITSKHASPDKWNDKPKNQLNKETDPVKSPPLPEHQKIPCNSAEPKSIPALSMETQKLGSVLSPESPKPTPLTPLEPQKPGSVVSPELQTPLPSPEPSKPASVSSPEPP |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: C13orf8 CAMPchromosome 13 open reading frame 8 Chromosome alignment-maintaining phosphoprotein FLJ90413 KIAA1802 zinc finger protein 828; Chromosome alignment-maintaining phosphoprotein 1; Zinc finger protein 828
Gene Aliases: C13orf8; CAMP; CHAMP; CHAMP1; KIAA1802; MRD40; ZNF828
UniProt ID: (Human) Q96JM3
Entrez Gene ID: (Human) 283489
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.