Novus Biologicals
Manufacturer Code:NBP2843660.1ML
Catalog # NB032294
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | The immunogen is a synthetic peptide directed towards the middle region of human ZNF823 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. Aliquot and store at -20°:C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: HSZFP36 ZFP 36 for a zinc finger protein ZFP36 ZFP-36 for a zinc finger protein zinc finger protein 823 zinc finger protein ZFP-36; ZFP 36 for a zinc finger protein; Zinc finger protein 823; Zinc finger protein ZFP-36
Gene Aliases: HSZFP36; ZFP36; ZNF823
UniProt ID: (Human) P16415
Entrez Gene ID: (Human) 55552
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.