Invitrogen
Manufacturer Code:PA541222
Catalog # PIPA541222
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the middle region of human ZNF690 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: KOX31-like zinc finger protein; sim to KOX31-like Zfp; zinc finger and SCAN domain containing 29; zinc finger and SCAN domain containing protein 29; Zinc finger and SCAN domain-containing protein 29; Zinc finger protein 690
Gene Aliases: AW125441; RGD1559150; Zfp690; ZNF690; ZSCAN29
UniProt ID: (Human) Q8IWY8
Entrez Gene ID: (Horse) 100056404, (Dog) 100856740, (Human) 146050, (Pig) 100157803, (Rabbit) 100328678, (Mouse) 99334, (Rat) 311353
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.