Novus Biologicals
Manufacturer Code:NBP2865410.1ML
Catalog # NB034165
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | The immunogen is a synthetic peptide directed towards the middle region of human ZNF644 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. Aliquot and store at -20°:C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: BM-005 KIAA1221Zep-2 MGC60165 MGC70410 ZEP2 zinc finger motif enhancer binding protein 2 Zinc finger motif enhancer-binding protein 2 zinc finger protein 644; Zep-2; zinc finger motif enhancer binding protein 2; Zinc finger motif enhancer-binding protein 2; Zinc finger protein 644
Gene Aliases: BM-005; KIAA1221; MYP21; NatF; ZEP-2; ZEP2; ZNF644
UniProt ID: (Human) Q6PI06
Entrez Gene ID: (Human) 84146
Molecular Function: KRAB box transcription factor transcription factor zinc finger transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.