Invitrogen
Manufacturer Code:PA557815
Catalog # PIPA557815
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human ZNF638 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CTCL tumor antigen se33-1; CTCL-associated antigen se33-1; Cutaneous T-cell lymphoma-associated antigen se33-1; NP220 nuclear protein; Nuclear protein 220; Zinc finger matrin-like protein; Zinc finger protein 638; Zinc finger protein 638 Cutaneous T-cell lymphoma-associated antigen se33-1 CTCL-associated antigen se33-1 Nuclear protein 220 Zinc finger matrin-like protein NP220 ZFML
Gene Aliases: NP220; ZFML; Zfp638; ZNF638
UniProt ID: (Human) Q14966
Entrez Gene ID: (Human) 27332
Molecular Function:
DNA binding protein
RNA binding protein
nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.