Novus Biologicals
Manufacturer Code:NBP29419120UL
Catalog # NB022940
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 1-300 of human ZNF638 (NP_001239542.1). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°:C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CTCL tumor antigen se33-1; CTCL tumor antigen se33-1 CTCL-associated antigen se33-1 Cutaneous T-cell lymphoma-associated antigen se33-1 DKFZp686P1231 MGC26130 MGC90196 NP220 nuclear protein NP220zinc finger matrin-like Nuclear protein 220 ZFML Zfp638 Zinc finger matrin-like protein zinc finger protein 638; CTCL-associated antigen se33-1; Cutaneous T-cell lymphoma-associated antigen se33-1; NP220 nuclear protein; Nuclear protein 220; Zinc finger matrin-like protein; Zinc finger protein 638
Gene Aliases: NP220; ZFML; Zfp638; ZNF638
UniProt ID: (Human) Q14966
Entrez Gene ID: (Human) 27332
Molecular Function:
DNA binding protein
RNA binding protein
nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.