Novus Biologicals
Manufacturer Code:NB10074657
Catalog # NB10074657
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | The immunogen recognized by this antibody maps to a region between residue 200 and 250 of human zinc finger protein 592 using the numbering given in entry NP_055445.2 (GeneID 9640). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C. Do not freeze. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CAMOS KIAA0211MGC138437 MGC138439 SCAR5 spinocerebellar ataxia autosomal recessive 5 zinc finger protein 592; Zinc finger protein 592
Gene Aliases: CAMOS; KIAA0211; SCAR5; ZNF592
UniProt ID: (Human) Q92610
Entrez Gene ID: (Human) 9640
Molecular Function: KRAB box transcription factor transcription factor zinc finger transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.