Novus Biologicals
Manufacturer Code:NBP184883
Catalog # NBP184883
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | This antibody was developed against Recombinant Protein corresponding to amino acids:KQDLVKLDINGLPYGLCAGCVNLSKSASPGINVPPGTNRPGLGQNENLSAIEGKGKVGGLKTRCSSCNVKFESE |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Early hematopoietic zinc finger protein; EHZF Evi3 LIP3 ZFP521 zinc finger protein 263 Zinc finger protein FPM315 Zinc finger protein with KRAB and SCAN domains 12; LYST-interacting protein 3; Zinc finger protein 521
Gene Aliases: EHZF; Evi3; LIP3; ZNF521
UniProt ID: (Human) Q96K83
Entrez Gene ID: (Human) 25925
Molecular Function:
KRAB box transcription factor
transcription factor
zinc finger transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.