Novus Biologicals
Manufacturer Code:NBP232131
Catalog # NBP232131
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunoprecipitation (IP) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | The immunogen this antibody was made to maps to a region between residue 175 to 225 of human Zinc Finger Protein 503 using the numbering given in entry NP_116161.2 (GeneID 84858). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C. Do not freeze. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: KOX17Zinc finger protein 191 lacks zinc finger domain Retinoic acid suppression protein A RSG-A Zfp191 Zinc finger and SCAN domain-containing protein 3 zinc finger protein 24 zinc finger protein 24 (KOX 17) ZNF191 ZSCAN3Zinc finger protein KOX17; Zinc finger protein 503
Gene Aliases: Nlz2; NOLZ-1; NOLZ1; ZNF503
UniProt ID: (Human) Q96F45
Entrez Gene ID: (Human) 84858
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.