Proteintech
Manufacturer Code:252281AP150UL
Catalog # 50-173-4801
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | ZNF451 Fusion Protein Ag18346 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Coactivator for steroid receptors; COASTER KIAA0576 KIAA1702 zinc finger protein 451 ZNF451; E3 SUMO-protein ligase ZNF451; E3 SUMO-protein transferase ZNF451; Zinc finger protein 451
Gene Aliases: COASTER; dJ417I1.1; KIAA0576; KIAA1702; ZNF451
UniProt ID: (Human) Q9Y4E5
Entrez Gene ID: (Human) 26036
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.