Abnova
Manufacturer Code:PAB8868
Catalog # 89-133-989
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to human ZNF335. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C. For long term storage store at -80°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against synthetic peptide of ZNF335.
For Research Use Only
Protein Aliases: NIF-1; NIF1; NIF2; NRC-interacting factor 1; Zinc finger protein 335; zinc-finger/leucine-zipper co-transducer NIF1
Gene Aliases: MCPH10; NIF-1; NIF1; NIF2; ZNF335
UniProt ID: (Human) Q9H4Z2
Entrez Gene ID: (Human) 63925
Molecular Function: KRAB box transcription factor transcription factor zinc finger transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.