Invitrogen
Manufacturer Code:PA541588
Catalog # PIPA541588
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the C-terminal of human ZNF312 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Fez family zinc finger protein 2; forebrain embryonic zinc finger; Forebrain embryonic zinc finger-like protein 2; forebrain embryonic zinc-finger like; testis tissue sperm-binding protein Li 80P; too few; zinc finger protein; Zinc finger protein 312; Zinc finger protein Fez-like; zinc-finger protein
Gene Aliases: AI451466; AI852056; cadps; FEZ; FEZF2; FEZL; FKSG36; foreheadin; SI:dZ6A2.1; SI:dZ6A2.7; TOF; ZFP312; zgc:92690; ZNF312
UniProt ID: (Bovine) Q2VWH6, (Human) Q8TBJ5, (Mouse) Q9ESP5
Entrez Gene ID: (Bovine) 613683, (Dog) 476568, (Human) 55079, (Rabbit) 100358877, (Horse) 100050147, (Mouse) 54713, (Rat) 305719, (Guinea pig) 100713168, (Zebrafish) 60634
Molecular Function:
DNA binding protein
centromere DNA-binding protein
nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.