Invitrogen
Manufacturer Code:PA540661
Catalog # PIPA540661
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the N-terminal of human ZNF312 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: FEZ family Zinc finger 2 FEZ TOF FEZL FKSG36 ZFP312 ZNF312; Fez family zinc finger protein 2; forebrain embryonic zinc finger; Forebrain embryonic zinc finger-like protein 2; testis tissue sperm-binding protein Li 80P; Zinc finger protein 312; Zinc finger protein Fez-like
Gene Aliases: AI451466; AI852056; FEZ; FEZF2; FEZL; FKSG36; TOF; ZFP312; ZNF312
UniProt ID: (Bovine) Q2VWH6, (Human) Q8TBJ5, (Mouse) Q9ESP5
Entrez Gene ID: (Bovine) 613683, (Dog) 476568, (Human) 55079, (Horse) 100050147, (Rabbit) 100358877, (Mouse) 54713, (Rat) 305719, (Guinea pig) 100713168
Molecular Function: DNA binding protein centromere DNA-binding protein nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.