Invitrogen
Manufacturer Code:PA5109228
Catalog # PIPA5109228
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Zebrafish |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant protein |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C Avoid Freeze/Thaw Cycles store in dark |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: AI451466 AI852056 BOS_20906 cadps Fez Fez family zinc finger protein 2 Fezf2 FEZL FKSG36 Forebrain embryonic zinc finger-like protein 2 foreheadin SI:dZ6A2.1 SI:dZ6A2.7 TOF Zfp312 zgc:92690 Zinc finger protein 312 Zinc finger protein Fez-like ZNF312; forebrain embryonic zinc finger-like protein 2; forebrain embryonic zinc-finger like; too few; zinc finger protein; zinc-finger protein
Gene Aliases: cadps; fez; fezl; foreheadin; SI:dZ6A2.1; SI:dZ6A2.7; tof; zgc:92690
Entrez Gene ID: (Zebrafish) 60634
Molecular Function:
DNA binding protein
centromere DNA-binding protein
nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.