Novus Biologicals
Manufacturer Code:NBP2843550.1ML
Catalog # NB032283
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | The immunogen is a synthetic peptide directed towards the C-terminal region of human ZNF312 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. Aliquot and store at -20°:C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: FEZ family zinc finger 2 fez family zinc finger protein 2 FEZLFEZ FKSG36 FLJ10142 Forebrain embryonic zinc finger-like protein 2 Zfp312 Zinc finger protein 312TOFZNF312ZFP312 Zinc finger protein Fez-like; Fez family zinc finger protein 2; Forebrain embryonic zinc finger-like protein 2; testis tissue sperm-binding protein Li 80P; Zinc finger protein 312; Zinc finger protein Fez-like
Gene Aliases: FEZ; FEZF2; FEZL; FKSG36; TOF; ZFP312; ZNF312
UniProt ID: (Human) Q8TBJ5
Entrez Gene ID: (Human) 55079
Molecular Function:
DNA binding protein
centromere DNA-binding protein
nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.