Novus Biologicals
Manufacturer Code:NBP29414620UL
Catalog # NB022850
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 1-270 of human ZNF268 (NP_001159353.1). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°:C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Zinc finger protein 268; zinc finger protein 3; zinc finger protein 446 Zinc finger protein with KRAB and SCAN domains 20 ZKSCAN20FLJ20626 ZSCAN30; Zinc finger protein HZF3
Gene Aliases: HZF3; ZNF268
UniProt ID: (Human) Q14587
Entrez Gene ID: (Human) 10795
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.