Novus Biologicals
Manufacturer Code:NBP222448G
Catalog # NBP222448G
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | LG1 |
Immunogen | murine ZPR1 protein |
Conjugate | DyLight 488 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C in the dark. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: ERP1 EZFIT-related protein 1 KIAA1396MGC150657 MGC150658 Z1971 zinc finger protein 471; Zinc finger protein 259; Zinc finger protein ZPR1
Gene Aliases: ZNF259; ZPR1
UniProt ID: (Human) O75312
Entrez Gene ID: (Human) 8882
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.