Proteintech
Manufacturer Code:127141AP150UL
Catalog # 50-172-8197
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | ZNF238 Fusion Protein Ag3406 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 58 kDa repressor protein; 58 kDa repressor protein C2H2 171 RP58 TAZ 1 TAZ1 Transcriptional repressor RP58 ZBTB18 ZFP238 zinc finger protein 238 Zinc finger protein C2H2 171 ZNF238; rRP58; TAZ-1; Transcriptional repressor RP58; Translin-associated zinc finger protein 1; Zfp-238; Zinc finger and BTB domain-containing protein 18; Zinc finger protein 238; Zinc finger protein C2H2-171
Gene Aliases: C2H2-171; MRD22; RP58; TAZ-1; TAZ1; ZBTB18; Zfp238; ZNF238
UniProt ID: (Human) Q99592, (Mouse) Q9WUK6, (Rat) Q9JKY3
Entrez Gene ID: (Human) 10472, (Mouse) 30928, (Rat) 64619
Molecular Function: DNA binding protein centromere DNA-binding protein nucleic acid binding transcription factor zinc finger transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.