Invitrogen
Manufacturer Code:PA5116935
Catalog # PIPA5116935
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant protein of human ZNF207 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 49 kDa zinc finger protein; 49 kDa zinc finger protein 8430401D15Rik BUB3-interacting and GLEBS motif-containing protein ZNF207 BUGZ hBuGZ Zep Zfp207 zinc finger protein 207 ZNF207; BUB3-interacting and GLEBS motif-containing protein ZNF207; BuGZ; Zinc finger protein 207
Gene Aliases: 8430401D15Rik; BUGZ; hBuGZ; Zep; Zfp207; ZNF207
UniProt ID: (Human) O43670, (Mouse) Q9JMD0
Entrez Gene ID: (Human) 7756, (Mouse) 22680, (Rat) 303763
Molecular Function:
DNA binding protein
nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.