Invitrogen
Manufacturer Code:PA553402
Catalog # PIPA553402
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human ZNF179 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: AI854096 R75574 ZNF179 Zfp179 bfp RING finger protein 112 brain finger protein zinc finger protein 179; Brain finger protein; neurolastin; RING finger protein 112; Zinc finger protein 179
Gene Aliases: BFP; RNF112; ZNF179
UniProt ID: (Human) Q9ULX5
Entrez Gene ID: (Human) 7732
Molecular Function:
G-protein
enzyme modulator
heterotrimeric G-protein
ligase
ubiquitin-protein ligase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.