Invitrogen
Manufacturer Code:PA531075
Catalog # PIPA531075
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide corresponding to a region within amino acids 585 and 604 of Mouse ZNF179 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: AI854096 R75574 ZNF179 Zfp179 bfp RING finger protein 112 brain finger protein zinc finger protein 179; Brain finger protein; Neurolastin; RING finger protein 112; Zinc finger protein 179
Gene Aliases: Bfp; Rnf112; Zfp179; ZNF179
UniProt ID: (Mouse) Q96DY5
Entrez Gene ID: (Mouse) 22671
Molecular Function: G-protein enzyme modulator heterotrimeric G-protein ligase ubiquitin-protein ligase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.