Novus Biologicals
Manufacturer Code:NBP293335100UL
Catalog # NB028554
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 200-500 of human RNF112 (NP_009079.2). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°:C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: BFPZinc finger protein 179ZNF179Brain finger protein FLJ99413 ring finger protein 112; Brain finger protein; neurolastin; RING finger protein 112; Zinc finger protein 179
Gene Aliases: BFP; RNF112; ZNF179
UniProt ID: (Human) Q9ULX5
Entrez Gene ID: (Human) 7732
Molecular Function:
G-protein
enzyme modulator
heterotrimeric G-protein
ligase
ubiquitin-protein ligase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.