Novus Biologicals
Manufacturer Code:NBP29423020UL
Catalog # NB023018
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 1-210 of human ZNF16 (NP_008889.2). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°:C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: KOX9Zinc finger protein KOX9 zinc finger protein 16 zinc finger protein 16 (KOX 9); Zinc finger protein 16; Zinc finger protein KOX9; zinc finger protein Kruppel type 9
Gene Aliases: HZF1; KOX9; ZNF16
UniProt ID: (Human) P17020
Entrez Gene ID: (Human) 7564
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.